missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tropomyosin beta chain Protein
A DNA sequence encoding the Homo sapiens (Human) Tropomyosin beta chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8763-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_003280.2 | |
Tropomyosin beta chain Protein | |
Research Use Only | |
TMSB | |
Recombinant Protein | |
E. coli | |
DKENAIDRAE QAEADKKQAE DRCKQLEEEQ QALQKKLKGT EDEVEKYSES VKEAQEKLEQ AEKKATDAEA DVASLNRRIQ LVEEELDRAQ ERLATALQKL EEAEKAADES ERGMKVIENR AMKDEEKMEL QEMQLKEAKH IAEDSDRKYE EVARKLVILE GELERSEERA EVAESKCGDL EEELKIVTNN LKSLEAQADK YSTKEDKYEE EIKLLEEKLK EAETRAEFAE RSVAKLEKTI DDLEDEVYAQ KMKYKAISEE LDNALNDITS L | |
Greater than 86.7% as determined by SDS-PAGE. |
7169 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction