missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Acetylcholinesterase/ACHE Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 5518-CE-010
301.50 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to cleave Acetylthiocholine. The specific activity is >250 nmol/min/μg, as measured under the described conditions.Specifications
P21836 | |
Supplied as a 0.2 μm filtered solution in Sodium Acetate and NaCl. | |
M.W. (Observed): 68-73 kDa, reducing conditions; M.W. (theoretical): 66 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
Ache | |
Recombinant | |
Acetylcholinesterase/ACHE |
Enzyme Activity | |
43 | |
Acetylcholinesterase/ACHE | |
Mouse myeloma cell line, NS0-derived mouse Acetylcholinesterase/ACHE protein Glu32-Leu614, with a C-terminal 10-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |