missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Complement Component C2 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 6725-SE-010

416.40 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18166330

  • 529.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to cleave a colorimetric peptide substrate, N-carbobenzyloxy-Gly-Arg-ThioBenzyl ester (Z-GR-SBzl), in the presence of 5,5’Dithio-bis (2-nitrobenzoic acid) (DTNB). Edwards, K.M. et al. (1999) J. Biol. Chem. 274:30468. The specific activity is >100 pmol/min/µg, as measured under the described conditions.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Sodium Acetate and NaCl.
58 kDa and 84 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
12263
Complement Component C2
Mouse myeloma cell line,NS0-derived mouse Complement Component C2 protein Met1-Leu760,with a C-terminal 6-His tag.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions