missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TWEAK/TNFSF12 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
529.00 €
Specifications
Gene ID (Entrez) | 21944 |
---|---|
Molecular Weight (g/mol) | 17 kDa |
Name | TWEAK/TNFSF12 |
Quantity | 25 μg |
Source | E. coli-derived mouse TWEAK/TNFSF12 protein Arg105-His249,with an N-terminal Met and a 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18106880
|
R&D Systems™
1237-TW-025 |
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. |
529.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18126140
|
R&D Systems™
1237-TW-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. |
529.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse TWEAK/TNFSF12 Protein is derived from E. coli. The Recombinant Mouse TWEAK/TNFSF12 Protein has been validated for the following applications: Bioactivity.Specifications
21944 | |
TWEAK/TNFSF12 | |
E. coli-derived mouse TWEAK/TNFSF12 protein Arg105-His249,with an N-terminal Met and a 6-His tag | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
17 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |