missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other MT II Protein

A cDNA sequence encoding the MT II was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12746-20mg

 View more versions of this product

Product Code. 15941409

  • 452.00 € / 20mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

MT II Protein
Research Use Only
Recombinant Protein
E. coli
The protein (1 mg/ml) was lyophilized with no additives.
20 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions