missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant P. multocida Hyaluronan Synthase/HAS Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 9585-GT-050
293.73 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant P. multocida Hyaluronan Synthase/HAS Protein is derived from E. coli. The Recombinant P. multocida Hyaluronan Synthase/HAS Protein has been validated for the following applications: Enzyme Activity.Specifications
Q7BLV3 | |
Supplied as a 0.2μm filtered solution in Tris and NaCl. | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
Enzyme Activity | |
81 kDa | |
E. coli-derived p. multocida Hyaluronan Synthase protein Met1-Ile703,with C-terminal 6-His tag | |
<0.10 EU per 1μg of the protein by the LAL method. | |
P. multocida |