missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MIF Protein
A cDNA sequence encoding the MIF was constructed and used to recombinantly synthesize the protein.
137.00 € - 5450.00 €
Specifications
Name | MIF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951259
|
enQuireBio™
QP12696-5ug |
5 μg |
137.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941259
|
enQuireBio™
QP12696-25ug |
25 μg |
212.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15931259
|
enQuireBio™
QP12696-1mg |
1 mg |
5450.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
MIF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4 and 5% trehalose. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPMFIVNTNV PRASVPEGFL SELTQQLAQA TGKPAQYIAV HVVPDQLMTF SGTSDPCALC SLHSIGKIGG AQNRNYSKLL CGLLSDRLHI SPDRVYINYY DMNAANVGWN GSTFA | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |