missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rubella Virus Rubella Capsid Protein
A cDNA sequence encoding the Rubella Capsid was constructed and used to recombinantly synthesize the protein.
238.00 € - 1515.00 €
Specifications
Name | Rubella Virus Rubella Capsid Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | ubella Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953359
|
enQuireBio™
QP13372-100ug |
100 μg |
238.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15973359
|
enQuireBio™
QP13372-500ug |
500 μg |
910.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15963359
|
enQuireBio™
QP13372-1mg |
1 mg |
1515.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Rubella Virus Rubella Capsid Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
ubella Virus | |
GST | |
Rubella protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl pH 8.0, 10mM glutation, 0.2% sarcosyl and 50% glycerol. |