Learn More
Bellingham Stanley™ Refractometer Calibration Oil
Product Code. 16299421
Click to view available options
:
±0.000074 RI
Product Code. 16299421
Brand: Bellingham Stanley™ 90531
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bellingham Stanley™ Refractometer Calibration Oils cover the range 1.47 -1.56 RI and are are ideal for calibrating refractometers that have temperature control in both food and industrial applications.
  • For refractometer calibration
  • A mix of organic compounds that cover the range 1.47 -1.56 RI at the top end of the Brix scale
  • Sold as a pack of five 5 mL glass bottles
  • Includes plastic pipette
  • 12-months shelf life
  • Traceable to NIST
  • Supplied with UKAS certificate of calibration
  • Value to five decimal places
TRUSTED_SUSTAINABILITY
Specifications
Type Calibration Oil
Refractive Index 1.53625
For Use With (Equipment) Refractometer
Accuracy ±0.000074 RI
Includes Dropping pipette
Quantity 5 x 5 mL