missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ Regenerated Cellulose Dialysis Tubes Membrane
Click to view available options
MWCO:
14 kDa
Unit Size:
30m
Specifications
Specifications
| Product Type | Dialysis Tubes Membrane |
| MWCO | 14 kDa |
| Material | Regenerated Cellulose |
| Diameter (Metric) | 6.3 mm |
| Length (Metric) | 30 m |
| Color | Colorless |
| Quantity | 1 Each |
| For Use With (Application) | Dialysis and ultrafiltration in the laboratory |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction