Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand:  Thermo Scientific™ R20412
57.48 EUR valid until 2025-04-30
Use promo code "24235" to get your promotional price.
 View more versions of this product
Product Code. 10413843
  • 60.50 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbidity Standard 2.0
15 x 103 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Instructions
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6212745 Certificate TypeCertificate of Analysis Date01/04/2025 Catalog NumberR20412
Lot Number6212744 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberR20412
Lot Number6185624 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR20412
Lot Number6175839 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog NumberR20412
Lot Number6155591 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog NumberR20412
Promotions
Promotions
  • Multiple Manufacturers
    Discover exclusive monthly offers on scientific equipment to enhance your laboratory's capabilities.
    Promo Code: 24235
    Expires2025-04-30
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards > Turbidity Standard 2.0