Learn More
Unity Lab Services™ Remote Monitoring Requalification Service
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Remote Monitoring Equipment
missing translation for 'unitSize'
1 unit
Description
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications and conforms to standards and requirements.
- Key tests include sensors and any accessories are correctly assembled and installed, and audit trails are reviewable and printable.
- Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
Specifications
Specifications
| For Use With (Equipment) | Remote Monitoring Equipment |
| Product Type | Requalification Service |