Learn More
Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes
Click to view available options
Needle Gauge:
18 G
Unit Size:
Pack of 5
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
| Material | Borosilicate Glass (Barrel) |
| Needle Gauge | 18 G |
| Diameter (Metric) Inner Needle | 0.80 mm |
| Diameter (Metric) Outer Needle | 1.27 mm |
| Product Type | Replacement Needles for Luer-Lock Syringes |
| Quantity | 5/Pk. |
| Length (Metric) Needle | 7 cm |
| Needle Point Style | Bevel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion