Learn More
Thermo Scientific™ Consumable Parts for Integrated Hydride Kit for iCAP™ ICP-OES Systems
Ensure peak performance of your ICP-OES analyzer with dedicated consumables for analysis of hydride-generating elements.
Brand:  Thermo Scientific™ 842318050151
 View more versions of this product
Product Code. 17691795
  • 330.65 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure peak performance of your Thermo Scientific™ iCAP™ Series ICP-OES system using dedicated Thermo Scientific™ Consumable Parts for the Integrated Hydride Kit for ICP-OES systems.
Peak performance with dedicated consumables
The replacement pump tubing and a consumables kit ensures the peak performance of hydride analysis on your iCAP Series ICP-OES System.
Consumables kit contains:
  • Pump tubing (for both, mini (95 mm) and standard peristaltic pump (152 mm))
  • Teflon membrane
  • O-ring
  • Replacement tubing
  • Sample probe
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Consumables Kit
Consumables Kit for Integrated Hydride Kit
Integrated Hydride Kit for ICP-OES