missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Reservoir Bottle
Click to view available options
Capacity (Metric):
2000 mL
Unit Size:
Each
Description
- For automatic burettes acc. to Pellet
- NS 29/32
- Country of origin is EC
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Color | Clear |
| Description | NS 29/32, country of origin is EC |
| Material | Glass |
| No. per Pack | 1 Each |
| Product Type | Reservoir Bottle |
| For Use With (Equipment) | For automatic burettes acc. to Pellet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction