Learn More
Fisherbrand™ Reverse Osmosis Membrane with Housing
Deionizes tap water and filters out particles for laboratory procedures. Fisher Scientific™ Reverse Osmosis Membrane with Housing is for use with the Accu500 Ultrapure Water System.
Brand: Fisherbrand™ RAR0100HG
This item is not returnable.
View return policy
Description
- For use with Accu500.
- Two sizes: 10L / 20L or 15L / 30L
- Removes particles, ions, and organics from tap water
Specifications
Accu500 10/20L | |
Reverse Osmosis Membrane | |
Housing |