missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ro 46-2005, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Ro 46-2005 is a novel synthetic non-peptide endothelin receptor antagonist, inhibits the specific binding of 125I-ET-1 to human vascular smooth muscle cells (ETA receptor) with IC50 of 220 nM.
Specifications
Specifications
| Chemical Name or Material | Ro 46-2005 |
| CAS | 150725-87-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H27N3O6S |
| Quantity | 10 mg |
| Solubility Information | DMSO : 100 mg/mL (211.18 mM; Need ultrasonic) |
| SMILES | O=S(C1=CC=C(C(C)(C)C)C=C1)(NC2=NC=NC(OCCO)=C2OC3=CC=CC(OC)=C3)=O |
| Molecular Weight (g/mol) | 473.54 |
| Formula Weight | 473.54 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction