missing translation for 'onlineSavingsMsg'
Learn More

Ro 6842262, >98%, Tocris Bioscience™

Product Code. 30000904
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent LPA1 antagonist

Potent LPA1 antagonist (IC50 = 25 nM). Exhibits >1,200-fold selectivity for LPA1 over LPA3. Attenuates NHLF human lung fibroblast cell proliferation and contraction in vitro. Also reduces plasma histamine levels in mouse LPA-challenge model. Orally bioavailable.

Chemical Identifiers

CAS 1396006-71-5
Molecular Formula C28H26N4O4
Molecular Weight (g/mol) 482.54
InChI Key PXQUHYSYFWQRMF-LJQANCHMSA-N
Synonym 1-4'-4-methyl-5-1r-1-phenylethoxy carbonylamino-1h-1,2,3-triazol-1-yl 1,1'-biphenyl-4-yl cyclopropanecarboxylic acid, 1-4'-4-methyl-5-r-1-phenyl-ethoxycarbonylamino-1,2,3 triazol-1-yl-biphenyl-4-yl-cyclopropanecarboxylic acid, 1-4'-4-methyl-5-r-1-phenyl-ethoxycarbonylamino 1,2,3 triazol-1-yl-biphenyl-4-yl-cyclopropanecarboxylic acid
PubChem CID 71265339
IUPAC Name 1-[4-[4-[4-methyl-5-[[(1R)-1-phenylethoxy]carbonylamino]triazol-1-yl]phenyl]phenyl]cyclopropane-1-carboxylic acid
SMILES CC1=C(N(N=N1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4(CC4)C(=O)O)NC(=O)OC(C)C5=CC=CC=C5

Specifications

Linear Formula C28H26N4O4
Solubility Information Soluble to 100mM in DMSO and to 20mM in ethanol
Quantity 10 mg
Formula Weight 482.53
Chemical Name or Material 1-[4′-[4-Methyl-5-[[[(1R)-1-phenylethoxy]carbonylamino]-1H-1,2,3-triazol-1-yl][1,1′-biph
Grade >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.