missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RosetteSep™ Human CD45 Depletion Cocktail
Click to view available options
Quantity:
200 mL whole blood
40 mL whole blood
Unit Size:
1 set
Description
- The RosetteSep™ Human CD45 Depletion Cocktail is designed to enrich epithelial circulating tumor cells (CTCs) from whole blood by depleting CD45+ cells
- Unwanted cells are targeted for depletion with Tetrameric Antibody Complexes recognizing CD45, CD66b and glycophorin A on red blood cells (RBCs)
- When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
- The purified epithelial tumor cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium
Specifications
Specifications
| Cell Type | Cancer Cells and Cell Lines |
| For Use With (Application) | Cell Isolation |
| Quantity | 40 mL whole blood |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction