missing translation for 'onlineSavingsMsg'
Learn More

Carl Roth™ Roti Colloidal Coomassie Staining Solution

Product Code. 10250132
Encompass
Click to view available options
Quantity:
Each
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 10250132

Brand: Carl Roth™ A152.1

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Specifications

Color Blue
Concentration 5X
Product Type Protein Stain
Stain Type Colloidal Coomassie Brilliant Blue
Form Liquid
For Use With (Application) For coomassie brilliant blue G250 staining agent for proteins in Polyacrylamide gels
Material Colloidal Comassie Brilliant Blue
Protein Load Yes
Quantity Each
Storage Requirements 20°C
Sufficient For 50 Mini Gels
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.