Learn More
Rotronic™ Disposable Sample Cups
Click to view available options
Depth (English):
1.57 in.
Depth (Metric):
4 cm
Unit Size:
Pack of 100
Description
- Prevent sample holders from coming into direct contact with product being tested, thereby preventing soiling or cross contamination
- Provides convenient means of collecting and storing samples
Specifications
Specifications
| Depth (Metric) | 4 cm |
| Depth (English) | 1.57 in. |
| Product Type | Sample Cup |
| Disposable | Single-use |
| For Use With (Application) | Collecting and Storing samples |
| For Use With (Equipment) | WP-40 or WP-40TH sample holders |