Learn More
Hach Lange™ Round Glass Disposable Cuvettes
Use for spectral measurements of liquids. Hach Lange™ Round Glass Disposable Cuvettes are important accessories for spectrophotometers.
Brand: Hach Lange™ LYY621
Description
- Round
- Disposable
- 50x10 mm
- 50 pcs
- Glass
Specifications
Cuvette | |
Glass | |
10 mm | |
Spectral Measurement of Liquids |
Single-use | |
560Pack | |
For LICO Spectrophotometers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hach Lange™ Round Glass Disposable Cuvettes > Path Length:11mm
Spot an opportunity for improvement?Share a Content Correction