missing translation for 'onlineSavingsMsg'
Learn More

RS 102221 hydrochloride, Tocris Bioscience™

Product Code. 30002151
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective 5-HT2C antagonist

Potent and selective 5-HT2C antagonist (pKi = 8.7). Displays ∽ 100-fold selectivity over the 5-HT2A and 5-HT2B subtypes and is > 100-fold selective over other 5-HT receptors, α- and β-adrenergic and muscarinic ACh receptors.

Chemical Identifiers

CAS 187397-18-8
Molecular Formula C27H32ClF3N4O7S
Molecular Weight (g/mol) 649.079
InChI Key SZJZEJZLRDIHPB-UHFFFAOYSA-N
Synonym rs 102221 hydrochloride, 8-5-2,4-dimethoxy-5-4-trifluoromethylphenylsulphonamido phenyl-5-oxopentyl-1,3,8-triazaspiro 4.5 decane-2,4-dione hydrochloride
PubChem CID 18931299
IUPAC Name N-[5-[5-(2,4-dioxo-1,3,8-triazaspiro[4.5]decan-8-yl)pentanoyl]-2,4-dimethoxyphenyl]-4-(trifluoromethyl)benzenesulfonamide;hydrochloride
SMILES COC1=CC(=C(C=C1C(=O)CCCCN2CCC3(CC2)C(=O)NC(=O)N3)NS(=O)(=O)C4=CC=C(C=C4)C(F)(F)F)OC.Cl

Specifications

Quantity 50 mg
Formula Weight 649.08
Percent Purity >98%
Chemical Name or Material RS 102221 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'