Learn More
(S)-(+)-2-Phenylglycine methyl ester hydrochloride, 97%, Thermo Scientific Chemicals
30.20 € - 97.40 €
Chemical Identifiers
CAS | 15028-39-4 |
---|---|
Molecular Formula | C9H11NO2·ClH |
Molecular Weight (g/mol) | 201.65 |
InChI Key | DTHMTBUWTGVEFG-QRPNPIFTSA-N |
Synonym | h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride |
PubChem CID | 11217910 |
IUPAC Name | methyl (2S)-2-amino-2-phenylacetate;hydrochloride |
SMILES | COC(=O)C(C1=CC=CC=C1)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12337153
|
Thermo Scientific Acros
442190100 |
10 g |
30.20 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12347153
|
Thermo Scientific Acros
442190500 |
50 g |
97.40 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15028-39-4 | |
201.65 | |
h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride | |
methyl (2S)-2-amino-2-phenylacetate;hydrochloride |
C9H11NO2·ClH | |
DTHMTBUWTGVEFG-QRPNPIFTSA-N | |
11217910 | |
COC(=O)C(C1=CC=CC=C1)N.Cl |
Specifications
15028-39-4 | |
96% min. (HPLC) | |
C9H11NO2·ClH | |
DTHMTBUWTGVEFG-QRPNPIFTSA-N | |
methyl (2S)-2-amino-2-phenylacetate;hydrochloride | |
11217910 | |
97% |
200°C | |
Glass Bottle | |
h-phg-ome.hcl, s-methyl 2-amino-2-phenylacetate hydrochloride, s-+-2-phenylglycine methyl ester hydrochloride, s-2-phenylglycine methyl ester hydrochloride, aminophenylacetic acid methyl ester, h-phg-ome hydrochloride, h-phg-ome hcl, d---alpha-phenylglycine methyl ester hydrochloride, methyl 2s-2-amino-2-phenylacetate hydrochloride, d-phenylglycine methyl ester hydrochloride | |
COC(=O)C(C1=CC=CC=C1)N.Cl | |
201.65 | |
201.65 | |
(S)-(+)-2-Phenylglycine methyl ester hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
- (S)-(+)-2-Phenylglycine methyl ester hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only