Learn More
Vötschtechnik™ Simpati™ Control Software
Optimize your environmental testing systems for research, development, production and quality assurance with this test system integration software package. Simpati™ Control Software offers easy, time saving operation for evaluation and documentation of test cycles and production sequences.
Brand: Vötschtechnik™ 64241243
Description
- Control of test and process cycles
- Convenient evaluation due to clear graphics and calculation options
- Administration of various users
- Networking of up to 99 test systems with one PC
- Program management for production cycles and tests
- Easy creation of test programs with the graphic, symbolic or shock test editor
- Loading of programs, production data via barcode
Options
- Loading of programs, production data (product, user, system) via barcode
- Monitoring of test systems in Internet Explorer 7.0 via ASP.NET application to Internet information Server (IIS 6.0) in Windows 2003 Server / 2008
- External module enables electronic signature on electronic paperless test documentation
- Easy-to-use interface EBO
- Acoustic error signal for Part 11 enabling (S!MPATI*-Pharma)
Specifications
Evaluation and documentation of test cycles and production sequences as well as the integration of special measuring data guarantees a higher quality standard. | |
Vötsch Environmental simulation and Heat technology product lines |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.