Learn More
MedChemExpress (S,R,S)-AHPC-PEG4-NH2
MedChemExpress (S,R,S)-AHPC-PEG4-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and 4-unit PEG linker used in PROTAC technology.
Brand: MedChemExpress HY-103604A
Specifications
663.83 g/mol | |
Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month | |
500 mg | |
DMSO : 170 mg/mL (256.09 mM; Need ultrasonic) |
2010159-57-4 | |
Room temperature | |
VH032-PEG4-NH2 ∣ VHL Ligand-Linker Conjugates 4 ∣ E3 ligase Ligand-Linker Conjugates 7 Free Base |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.