missing translation for 'onlineSavingsMsg'
Learn More

S100A4 Mouse anti-Human, Mouse, Clone: 2G11B4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66489-1-IG-150UL

 View more versions of this product

Product Code. 16818415

  • 410.00 € / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
Specifications

Specifications

S100A4
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P07091, P26447
S100A4
S100A4 Fusion Protein ag9019
150 μL
Primary
Human, Mouse
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2G11B4
Unconjugated
S100A4
18A2, 42A, Calvasculin, CAPL, Fibroblast-specific protein 1, FSP1, FSP-1, Metastasin, MTS1, P9KA, PEL98, Protein Mts1, Protein S100 A4, S100A4
Mouse
Protein A
RUO
20198, 6275
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.