missing translation for 'onlineSavingsMsg'
Learn More

S100B Mouse, Clone: 4C4.9, enQuireBio™

Mouse Monoclonal Antibody

199.00 € - 510.00 €

Specifications

Antigen S100B
Clone 4C4.9
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15940838
View Documents Promotion Details
EnquireBio
6285-MSM1-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15950838
View Documents Promotion Details
EnquireBio
6285-MSM1-P1
100 μg
510.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

S100B
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
6285
Purified bovine brain S100 protein
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
10-12kDa
4C4.9
Immunohistochemistry
Unconjugated
RUO
XP_016883913
S100B
IgG2a κ
Protein A/G Purified
Cytoplasmic and Nuclear
Brain, Melanoma
Videos
SDS
Documents

Documents

Product Certifications