Learn More
Greiner Bio-One Sapphire PCR Tube without Cap
For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tube without Cap are comprehensive thermal cycler compatible.
Brand: Greiner Bio-One 684201
This item is not returnable.
View return policy
Description
- Thin-walled for heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Packaging labelled with product symbols for warehousing
- Used for sample handling
Specifications
Natural | |
PCR | |
Polypropylene |
0.2 mL | |
Non-sterile | |
PCR Tube without Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One Sapphire PCR Tube without Cap > Sapphire PCR Tube without Cap
Spot an opportunity for improvement?Share a Content Correction