Learn More
Greiner Bio-One Sapphire PCR Tube without Cap
For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tube without Cap are comprehensive thermal cycler compatible.
Brand:  Greiner Bio-One 684201
Product Code. 10155151
  • 85.11 € / Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Natural
PCR
Polypropylene
0.2 mL
Non-sterile
PCR Tube without Cap
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One Sapphire PCR Tube without Cap > Sapphire PCR Tube without Cap