Learn More
Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap
Product Code. 10247111
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Volume (Metric):
0.2 mL
0.5 mL
Unit Size:
Pack of 1000
Pack of 10000
This item is not returnable. View return policy
Product Code. 10247111
Brand: Greiner Bio-One 683201
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
For use in RT/Quantitative-PCR. Greiner Bio-One Sapphire PCR Tubes with Attached Frosted Flat Cap are comprehensive thermal cycler compatible.
  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Volume (Metric) 0.2 mL
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap