Learn More
Sartorius™ Sartobind™ Polypropylene Housing Stabilized Reinforced Cellulose Membrane Q SingleSep Nano Capsules
Product Code. 11770316 Shop All Sartorius Products
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. 11770316
Brand: Sartorius™ 92IEXQ42DN11
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance USP Class VI
Material (Membrane) Anion Exchanger Stabilized Reinforced Cellulose
Pore Size 3 to 5 μm
Filtration Area 36 cm2
For Use With (Application) For DNA, host cell proteins, endotoxins, viruses contaminant removal applications
Housing Material Polypropylene
No. per Pack 1Each
Quantity 1 Each
Type Q SingleSep Nano Capsules