Learn More
Sartorius arium™ Foot Switch
More Convenient Pure Water Dispensing
Brand:  Sartorius H2O-AFS1
Product Code. 11754996
  • 569.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For external control of the time / volume dispense port for arium 611 systems.
  • Water dispensing at a press of the foot
  • Ideally suited for clean rooms, minimize the risk of contamination
  • Comfortable fatigue-free switching thanks to low height
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Arium Water Purification System
Foot Switch