Learn More
Sartorius arium™ Tank Level Switch
For external control of an open gravity tank for arium™ pro systems. The arium™ Tank Level Switch is a system accessory to assist with daily use of arium™ lab water systems.
Brand:  Sartorius H2O-ALS1
Product Code. 11764996
  • 703.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The level sensor makes it easy to connect an external water storage tank and subsequently fill a tank with ultrapure water.
  • Used with the arium™ multifunction box
  • Flexible water transport to anywhere
  • Can fill every tank system
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Water System
Level Sensor
3 m Cable
2.03 cm (bore diameter: 1.65 cm)