Learn More
Sartorius Replacement Part for Polycarbonate Filtration Holders
Replacement Part for Polycarbonate Filtration Holders is for use with Microsart™ Manifolds 16510 and 16511.
Brand: Sartorius 6981090
25.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polycarbonate Accessory
- For use with Microsart™ Manifolds 16510, 16511
Specifications
Reusable | |
Polycarbonate | |
1 Ea. |
Filter Holder | |
Non-sterile | |
Receiver Flask |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius Replacement Part for Polycarbonate Filtration Holders > Replacement Part for Polycarbonate Filtration Holders; Type: Receiver Flask; Quantity: 1 Ea.
Spot an opportunity for improvement?Share a Content Correction