Learn More
Sartorius Sartolab™ 150v Vacuum Filtration Units
Shop All Sartorius ProductsDescription
Sartolab 150v is a disposable, 0.2μm sterile, ready-to-use membrane filter capsule for highest convenience. Sartolab 150v capsules are made of a unique hydrophilic Polyethersulfone (PES) membrane providing outstanding total throughput, flow rate performance, low extractables and broadest chemical compatibility. Each 0.2 ++m bottle-top capsule filters 15 liters of aqueous solutions. The integrated on-off valve switch allows easy movement between receiving flasks.
- Quality and reliability
- Low protein binding
- Fast flow and high throughput
- Easy to use
Specifications
Specifications
| Product Type | Flask Rack |
| Autoclavable | Not Autoclavable |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.