Learn More
Sartorius™ Vivapure™ D Mini H IEX Spin Column
Ready-to-use centrifugal devices based on Sartobind Membrane Adsorber technology as their chromatography matrix
Brand: Sartorius™ VS-IX01DH24
Additional Details : Weight : 0.75000kg
Description
Vivapure Mini Ion Exchange (IEX) spin columns are centrifugal devices, incorporating Sartobind Membrane Adsorber technology as their chromatography matrix. Vivapure IEX spin columns make protein purification as easy as filtration. The devices are ready-to-use and do not bear the risk of running dry. For many protein purification applications, they can replace time-consuming and tedious column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many different samples are processed in parallel. VIvapure Mini is available in Diethylamine (D) membrane chemistry.
Typical Applications
- Sample preparation prior to 1-D or 2-D PAGE
- Purification scouting of an unknown protein
- Removal of endotoxins from monoclonal antibodies
- Preparation of heme moiety from heme containing protein for functional analysis
- Polishing of His-tagged proteins
- General protein purification
- Detergent removal from protein solutions
- Purification of antibodies from serum, ascites or tissue culture supernatant
- Establishing a purification/ pre-purification protocol for a given protein
- HPLC/FPLC sample preparation
- Purification of membrane-bound proteins
- Fast centrifugal spin columns
- Easy and ready-to-use
- Binding capacity 4mg protein
- Ideal for parallel processing
Specifications
240 μL | |
High | |
3 to 5 μm | |
24/Pk. | |
Spin Column |
0.4 mL | |
Polypropylene | |
Stabilized regenerated Cellulose | |
Paralell processing |