Learn More
Fisherbrand™ Screw Caps
Product Code. 10696163
Click to view available options
Closure Size:
GL 25
GL 32
This item is not returnable. View return policy
This item is not returnable. View return policy
An excellent option for closing sample storage bottles in the laboratory.
  • With PTFE-coated silicone gasket
  • Acc. to DIN 168
  • Leak-proof with excellent resistance
Specifications
Material PBT
For Use With (Equipment) Screw-neck sample storage bottles
Closure Liner Silicone Gasket
Closure Size GL 25