missing translation for 'onlineSavingsMsg'
Learn More

Carl Roth™ Sekuroka™ Lens Cleaning Tissues

Product Code. 10142262
Encompass_Preferred
Click to view available options
For Use With (Application):
Delicate, Moist Cleaning of Optical Glasses
Description:
Wipe
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10142262

Brand: Carl Roth™ 1678.1

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Cleans glasses without leaving specks. Roth Carl™ Sekuroka™ Lens Cleaning Tissues for moist cleaning of optical glasses, with active grease remover and alcohol.

  • Anti-static and non-fogging
  • Only one wipe required
  • Individally packed

Specifications

Description Wipe
Product Type Cleaning Wipe
For Use With (Application) Delicate, Moist Cleaning of Optical Glasses
No. per Pack 100/Pk.
compliance-icons compliance-icons
Product Identifier
  • Lens Cleansing Tissues
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : ethyl alcohol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.