missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ Sekuroka™ Plastic Made Security Boxes
Click to view available options
Color: ; Dimensions (D x W x H): 120 x 236 x 120mm
Specifications
Specifications
Product Type | Security Boxes |
For Use With (Application) | Used to Protect Goggles, Ear Protector from Dust and Water |
No. per Pack | 1 Ea. |
Material | Plastic |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'