missing translation for 'onlineSavingsMsg'
Learn More

Semaglutide, MedChemExpress

Product Code. p-200096671
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16487527

Brand: Medchem Express HY114118

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Semaglutide, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide has the potential for type 2 diabetes treatment.

Semaglutide, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide has the potential for type 2 diabetes treatment.

Specifications

Chemical Name or Material Semaglutide
Physical Form Solid
CAS 910463-68-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C187H291N45O59
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 33.33 mg/mL (8.10 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (3.04 mM; ultrasonic and adjust pH to 2 with HCl)
SMILES [Semaglutide]
Molecular Weight (g/mol) 4113.64
Formula Weight 4113.64
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.