missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Senataxin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
237.00 € - 558.00 €
Specifications
| Antigen | Senataxin |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18728133
|
Novus Biologicals
NBP1-94712SS |
0.025 mL |
237.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18761614
|
Novus Biologicals
NBP1-94712 |
558.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Senataxin Polyclonal specifically detects Senataxin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Senataxin | |
| Western Blot 1:500, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| DNA Repair | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
| SETX | |
| IgG | |
| Affinity Purified |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q7Z333 | |
| 23064 | |
| A genomic peptide made to an internal portion of the human Senataxin protein (between residues 600-750) [UniProt Q7Z333] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title