Learn More
Trajan™ Diamond GC Needles for 10 to 50μL Manual Syringes
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond GC Needles for 10 to 50μL Manual Syringes are MS ready and designed to improve the sensitivity of your MS results.
Brand: Trajan™ 0355310
This item is not returnable.
View return policy
Description
Improve the sensitivity of your MS analysis:
- Lowest syringe carryover
- Reduced non-specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
26 G | |
0.018 in. | |
0.45 mm | |
2 in. | |
Bevel |
0.0043 in. | |
0.11 mm | |
Ultra Smooth Hydrophilic Coating | |
GC Needle | |
Manual Syringe (10 to 50μL) |