Learn More
Trajan™ Diamond MS Autosampler Syringes: For Agilent
Product Code. p-4682046
Click to view available options
Volume (Metric):
10 μL
25 μL
Model:
10R-AG-GT-MS1
25R-AG-GT-MS1
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 13147976
Brand: Trajan™ 002760
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
For Use With (Application) Improves mass spectrometry analysis sensitivity
Product Type Diamond MS Syringe
Model 10R-AG-GT-MS1
Quantity Ea.
Volume (Metric) 10 μL
Connection Type Removable