Learn More
SGE™ BP20 (Wax) GC Capillary Columns
Product Code. 12915591
Click to view available options
Length (Metric):
10 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
2 μm
Unit Size:
Each
Product Code. 12915591
Brand: SGE™ 054405
Please to purchase this item. Need a web account? Register with us today!
Combines innovative phase technology and unique fused silica production capability. SGE™ BP20 (Wax) GC Capillary Columns provides end-to-end separation solutions for all applications.
Features
  • Industry standard Wax column
  • Bonded and cross-linked
  • Can be solvent rinsed
Applications
  • Used for separation of Alcohol, Free acids, Fatty acids methyl esters, Aromatics, Solvents and Essential oils
Specifications
Max. Temperature 280°C
Functionality/Interaction Type Hydrogen Bonding or Dipole interactions
Diameter (Metric) Inner 0.1 mm
Stationary Phase SGE™ BP20 (Wax)
USP Type G14, G15, G16, G20, G39
For Use With (Application) Solvents and Essential Oils Analysis, Free Acids, Fatty Acid Methyl Esters, Aromatics Analysis, Solvents and Essential Oils
Temperature Range 20°C to 260/280°C
Film Thickness 0.1 μm
Length (Metric) 10 m
Polarity Polar
Coating Fused Silica