Learn More
Epredia™ Cytospin™ Collection Fluid
Product Code. 12638016 Shop All Epredia Products
Click to view available options
Quantity:
120mL
500mL
4 L
Unit Size:
3.8 L
Pack of 125
Pack of 2
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 12638016
Brand: Epredia™ 9990323
Please to purchase this item. Need a web account? Register with us today!
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
  • Packaged in convenient pump bottle or pre-filled specimen cups
  • Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Collection Fluid
Quantity 120mL
Solution Type Alcohol/Carbowax (PEG) combination
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cytospin Collection Fluid
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Flammable liquid Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H350-May cause cancer.
  • H372-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : FD&C yellow No. 5, ethanol, isopropanol,,ethanol, Polyethylene glycol, Methylene blue trihydrate