missing translation for 'onlineSavingsMsg'
Learn More

SHOWA A0170 Anti-static Knit Gloves

Product Code. 17821201
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

The A0170 anti-static glove is suitable for handling glass, abrasive, and electrical components. It has a liner that dissipates static electricity and a polyurethane coating on the palm.

  • The A0170 antistatic glove has a polyurethane coating on the palm that protects against oils and abrasion.
  • The polyurethane coating also leaves no marks or fingerprints on handled objects.
  • The glove is designed for wearer comfort with a flexible, seamless liner.
  • The elasticated cuff of the glove prevents debris from entering.

Specifications

Chlorinated No
Construction Seamless Knit
Accelerator Free Yes
Applications Industrial, Laboratory
Coating Material Polyurethane
Color White, Gray
Disposable Reusable
Exterior Finish Partially Textured
Hand Specific
Laboratory/Cleanroom Use Cleanroom
Latex Free Yes
Material Nylon, Polyester, Polyurethane
Powdered Powder Free
Product Type Anti-static Gloves
Size Medium
Sterility Non-sterile
EN388 Standard EN388:3121X
Gender Unisex
Glove Cuff Style Knit
Tear Rating 2
Yarn Gauge 13 G
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.