missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-G Antibody (805903) [DyLight 680], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7775Y
This item is not returnable.
View return policy
Description
Siglec-G Monoclonal antibody specifically detects Siglec-G in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Siglec-G | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
89790 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
805903 | |
Flow Cytometry, CyTOF-ready | |
PRO940;Sialic acid-binding Ig-like lectin 10;SIGLEC10;SIGLEC-10;SLG2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Siglec-G, Glu20-Gln525, Accession # NP_766488 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |