Learn More
Ambion™ Silencer™ Custom Designed siRNA
Click to view available options
Purity:
HPLC
Standard
Quantity:
20 nmol
40 nmol
5 nmol
Dimensione della confezione:
20nmol
40nmol
5nmol
Descrizione
- Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
- Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
Specifica
Specifica
| Content And Storage | Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water. |
| Description | GeneAssist - Custom Designed Silencer™ siRNA |
| For Use With (Application) | Knocking down gene expression in order to study protein function in a wide range of cell types |
| Product Line | Silencer |
| Product Type | Custom Designed siRNA |
| Purity | Standard |
| Quantity | 5 nmol |
| Shipping Condition | Ambient |
| Temperature (Metric) | Upon receipt, store in a non-frost-free freezer at or below -20°C |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto