Learn More
Macherey-Nagel™ Silica 60 Adsorbent
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
1 kg
5 kg
Particle Size:
<0.08 mm
0.025 to 0.04 mm
0.05 to 0.2 mm
63 μm
Unit Size:
1kg
5kg
Specifications
Specifications
| Particle Size | 63 μm |
| Pore Size | 60 Å |
| Quantity | 1 kg |
| Type | Silica 60 |
| Material | Silica |