Learn More
Macherey-Nagel™ Silicone Standard Septa
Brand:  Macherey-Nagel™ 702610
 View more versions of this product
Product Code. 10025831
  • 47.70 € / Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Standard Septum
60° Shore A
50 Pack
10 mm
Silicone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Silicone Standard Septa > O.D.: 10mm